Specifications
- Antibody type:Primary
- Antigen name:Phosphodiesterase 4D
- Antigen symbol:PDE4D
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:5144
- Antigen synonyms:dunce-like phosphodiesterase E3|HSPDE4D|PDE4DN2|HGNC:8783|cAMP-specific phosphodiesterase 4D|phosphodiesterase 4D|phosphodie|PDE4D|Drosophila)|STRK1|cAMP-specific phosphodiesterase PDE4D6|DPDE3|cAMP-specific (phosphodiesterase E3 dunce homolog
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Cyclic AMP-dependent phosphodiesterase type D (PDE4D) family is comprise of 5 variants (PDE4D1, D2, D3, D4 and D5). One or more PDE4D subtype variants are ubiquitously present in all mammalian cells. In CNS all five PDE4D subtype variants are expressed in varying ratios and their activity is regulated in tandem with GPCRs stimulation. Peripheral tissues also exhibit differential expression of PDE4D variants. PDE4D1/D2 mRNA levels rise in response to an increase in cAMP. Short term regulation of PDE4D variants involved PKA, MAP kinases and Erk2 phosphorylation that results in rapid change in their enzymatic activities. Other regulatory mechanism involved protein protein interactions with cytoskeletal scaffolding proteins.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PDE4D
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat