- Antibody type:Primary
- Antigen name:FYN Binding Protein
- Antigen symbol:FYB
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:2533
- Antigen synonyms:SLAP130|SLAP-130|PRO0823|ADAP
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Fyb (Fyn binding protein) and the anchoring proteins SKAP55 (src kinase-associated phosphoprotein) and SKAP55-R (SKAP55-related protein) associate with the tyrosine kinase p59fyn (1–3). SKAP55 and SKAP55-R bind to Fyb through their SH3 domains and function as substrates for p59Fyn in resting T cells (1–3). SKAP55 contains an amino-terminal pleckstrin homology domain and a carboxy-terminal SH3 domain binding motif of adjacent arginine and lysine residues followed by tandem tyrosines (i.e. RKxxYxxY) (4,5). SKAP55-R, similar in overall structure to SKAP55, contains a coiled-coil N-terminal domain (1,2). SKAP55 associates with SLAP-130, another component of the Fyn complex, which plays a role in the regulation of signaling events initiated by lymphocyte antigen receptors leading up to T cell activation (6). The human Fyb gene maps to chromosome 5p13.1 and encodes a 783 amino acid protein (7).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: FYB/ADAP
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat