Order Entry
Switzerland
ContactUsLinkComponent
Anti-BCAR1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anti-BCAR1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-12574R-A488
Supplier:  Bioss
Anti-BCAR1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-12574R-A488
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Breast Cancer Anti-Estrogen Resistance 1
  • Antigen symbol:
    BCAR1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 488
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    9564
  • Antigen synonyms:
    Cas Scaffolding Protein Family Member 1|CASS1|CRK-Associated Substrate|Crk-Associated Substrate P130Cas|CRKAS
  • Modification:
    phospho-Tyr249
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

p130 represents one of several known substrates for v-Crk encoded p47. p130 Cas (for Crk-associated substrate) exhibits a high level of tyrosine phosphorylation and is tightly associated with v-Crk, suggesting a role in v-Crk-mediated cell signaling. The molecular cloning of p130 Cas has shown it to represent a novel SH3 containing signaling molecule with a cluster of multiple putative SH2-binding motifs for v-Crk. By immunoprecipitation analysis, p130 Cas has been shown to be highly phosphorylated at tyrosine residues subsequent to either v-Src p60 or v-Crk-mediated transformation and to form stable complexes with both of these transforming proteins. p130 Cas behaves as an extremely potent substrate for protein tyrosine kinases and has been reported to relocate from the cytoplasm to cell membrane upon tyrosine phosphorylation. One proposed model is that the SH2 domain of v-Crk functions to activate c-Src kinase, which in turn phosphorylates p130 Cas.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BCAR1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human