Specifications
- Antibody type:Primary
- Antigen name:Activin A Receptor Type 2A
- Antigen symbol:ACVR2A
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:92
- Antigen synonyms:Activin receptor type IIA|Activin receptor type-2A [Precursor]|ACTR-IIA|ACVR2|ACTRIIA
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Members of the transforming growth factor b superfamily bind to a pair of transmembrane proteins, known as receptor types I and II, which contain serine/threonine kinases and associate to form a signaling complex (1). Activin has been shown to bind a heteromeric noncovalent complex, which consists of a type I receptor, ACTR-IA (also designated ACVRI and ALK-2) or ACTR-IB (also designated ALK-4 and SKR2), and a type II receptor, ACTR-IIA (also designated ACVR2A) or ACTR-IIB (also designated ACVR2B) (1-6). Both receptor types are highly expressed in brain (5). The activin receptor family members are thought to mediate distinct effects on gene expression, cell differentiation, and morpho- genesis in a dose dependent fashion (5,6).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ACVR2A
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat