Order Entry
Switzerland
ContactUsLinkComponent
Anti-SOX6 Rabbit Polyclonal Antibody (Cy3®)
Anti-SOX6 Rabbit Polyclonal Antibody (Cy3®)
  BOSSBS-11599R-CY3
 :  Bioss
Anti-SOX6 Rabbit Polyclonal Antibody (Cy3®)
  BOSSBS-11599R-CY3
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    SRY (sex determining region Y)-box 6
  • Antigen symbol:
    SOX6
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy3®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

Sox genes comprise a family of genes that are related to the mammalian sex determining gene SRY. These genes similarly contain sequences that encode for the HMG-box domain, which is responsible for the sequence-specific DNA-binding activity. Sox genes encode putative transcriptional regulators implicated in the decision of cell fates during development and the control of diverse developmental processes. The highly complex group of Sox genes cluster at least 40 different loci that rapidly diverged in various animal lineages. At present, 30 Sox genes have been identified. Members of this family have been shown to be conserved during evolution and to play key roles during animal development. Some are involved in human diseases, including sex reversal.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SOX6
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat