Order Entry
Switzerland
ContactUsLinkComponent
Anti-AACS Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-11383R-FITC
Anbieter:  Bioss
Anti-AACS Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-11383R-FITC
Anbieter:  Bioss

Technische Daten

  • Antibody type:
    Primary
  • Antigen name:
    Acetoacetyl-CoA Synthetase
  • Antigen symbol:
    AACS
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    65985
  • Antigen synonyms:
    Acyl-CoA synthetase family member 1|Protein sur-5 homolog|Acetoacetyl-CoA synthetase|AACS_HUMAN.|ACSF1
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Technische Daten

Über diesen Artikel

ACSF1 is a 672 amino acid protein belonging to the ATP-dependent AMP-binding enzyme family. Encoded by a gene that maps to human chromosome 12q24.31, ACSF1 is highly expressed in kidney, heart and brain, and shows similar neural expression as HMGCR (3-hydroxy-3-methylglutaryl-CoA reductase). Existing as three alternatively spliced isoforms, ACSF1 participates in ATP binding, ligase activity, acetoacetate-CoA ligase activity and nucleotide binding. The ACSF1 promoter is a known PPAR?target gene, with the nuclear receptor recruited to the ACSF1 promoter by direct interaction with stimulating protein-1 (Sp1). ACSF1 activates acetoacetate and is highly regulated by modulators that affect HMGCR and cholesterol biosynthesis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AACS
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat