Anti-TIMP2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
BOSSBS-0416R-A555
: Bioss
- Antibody type:Primary
- Antigen name:Tissue inhibitor of metalloproteinases 2
- Antigen symbol:TIMP2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:7077
- Antigen synonyms:tissue inhibitor of metalloproteinases 2|metalloproteinase inhibitor 2|TIMP-2|tissue inhibitor of metalloproteinase 2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:120-165/220
- Purification:Purified by Protein A
- Pk:100 µl
Complexes with metalloproteinases (such as collagenases) and irreversibly inactivates them by binding to their catalytic zinc cofactor. Known to act on MMP-1, MMP-2, MMP-3, MMP-7, MMP-8, MMP-9, MMP-1, MMP-13, MMP-14, MMP-15, MMP-16 and MMP-19.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TIMP2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat