ContactUsLinkComponent
J.T.Baker® BAKERBOND Speedisk® C18 PolarPlus®, SPE Columns
J.T.Baker® BAKERBOND Speedisk® C18 PolarPlus®, SPE Columns
Catalog # 8153-04
Supplier:  Avantor
undefined
J.T.Baker® BAKERBOND Speedisk® C18 PolarPlus®, SPE Columns
Catalog # 8153-04
Supplier:  Avantor
Specifications
  • Capacity:
    3 ml
  • Seq:
    02
  • Bed weight:
    50 mg
  • Phase:
    Octadecyl (C18) PolarPlus
  • Pk:
    1
Specifications
About this item
Ultra-clean, PP columns with BAKERBOND® HPLC media are engineered to save time, minimise work and final solvent volumes and improve performance. Run times are up to nine times faster than conventional columns. The built-in column guard filter and higher analyte concentration may reduce or eliminate pre-filtration and concentration steps.
  • Run times up to nine times faster than conventional SPE columns
  • Unique laminar configuration
  • Silica based
General applications: Non ionic, basic, non polar to polar analytes.
These SPE columns can be used in reversed phase mode.