Order Entry
Switzerland
ContactUsLinkComponent
Multi-parameter meters, bench, SevenExcellence™ S479 / S975-uMix
Multi-parameter meters, bench, SevenExcellence™ S479 / S975-uMix
Supplier:  Mettler - Toledo
Certificates
undefined
Multi-parameter meters, bench, SevenExcellence™ S479 / S975-uMix
Supplier:  Mettler - Toledo

Specifications

  • Catalog No:
  • METT30092989
  • METT30092988
  • Model:
  • S975-uMix
  • S479
  • pH range:
  • −2,000...+20,000
  • −2,000...+20,000
  • pH resolution:
  • 0,001 / 0,01 / 0,1
  • 0,001 / 0,01 / 0,1
  • pH accuracy:
  • ± 0,002
  • ± 0,002
  • pH calibration:
  • Max. 5 points, 8 pre-defined and 20 user-defined buffer groups
  • Max. 5 points, 8 pre-defined and 20 user-defined buffer groups
  • mV range:
  • ± 2000,0
  • ± 2000,0
  • mV resolution:
  • 0,1 / 1
  • 0,1 / 1
  • mV accuracy:
  • ± 0,1
  • ± 0,1
  • ISE range:
  • 0 - 999 999 mg/L (ppm)
  • 0 - 999 999 mg/L (ppm)
  • ISE resolution:
  • 0,001 - 1
  • ISE accuracy:
  • ±0,5%
  • ±0,5%
  • Conductivity range:
  • 0,001 µS/cm - 2000 mS/cm
  • 0,001 µS/cm - 2000 mS/cm
  • Conductivity resolution:
  • 0,001 - 1
  • 0,001 - 1
  • Conductivity accuracy:
  • ±0,5%
  • ±0,5%
  • Conductivity calibration:
  • 13 pre-defined and 20 user-defined standards
  • 13 pre-defined and 20 user-defined standards
  • Resistivity range:
  • 0,01 - 100,0 MOhm*cm
  • 0,01 - 100,0 MOhm*cm
  • Salinity range:
  • 0,00 - 80,00 psu
  • 0,00 - 80,00 psu
  • TDS range:
  • 0,001 mg/L - 1000 g/L
  • 0,001 mg/L - 1000 g/L
  • DO concentration:
  • 0,000 - 50 (optical); 0,000 - 99 mg/l (polarographic)
  • 0,000 - 50 (optical); 0,000 - 99 mg/l (polarographic)
  • DO concentration resolution:
  • 0,001 mg/L
  • 0,001 mg/L
  • DO saturation:
  • 0,0 - 500 % (optical); 0,0 - 600 % (polarographic)
  • 0,0 - 500 % (optical); 0,0 - 600 % (polarographic)
  • DO accuracy:
  • Optical: ±0,1 mg/L (from 0 - 8); ±0,2 mg/L (from 8 - 20); ±10% (from 20 - 50)
    Polarographic: ±0,5%
  • Optical: ±0,1 mg/L (from 0 - 8); ±0,2 mg/L (from 8 - 20); ±10% (from 20 - 50)
    Polarographic: ±0,5%
  • DO calibration:
  • 2-point
  • 2-point
  • DO barometric pressure range:
  • 500 - 1100 mbar
  • 500 - 1100 mbar
  • Temperature range:
  • −30,0...+130,0 °C
  • −30,0...+130,0 °C
  • Temperature resolution:
  • 0,1 °C
  • 0,1 °C
  • Temperature accuracy:
  • ± 0,1 °C
  • ± 0,1 °C
  • Temperature compensation:
  • Linear, non linear, off
  • Linear, non linear, off
  • Display:
  • 7 inch colour TFT
  • 7 inch colour TFT
  • Data logging capacity:
  • At least 20 000 data points, 250 analyses
  • At least 20 000 data points, 250 analyses
  • Outputs:
  • USB, Ethernet and RS232
  • USB, Ethernet and RS232
  • Reference temperature:
  • 20 / 25 °C
  • 20 / 25 °C
  • W×D×H:
  • 235×188×75 mm
  • 235×188×75 mm
  • Weight:
  • 1,12 kg
  • 1,12 kg

Specifications

Related Information

Expansion units

Product Information

Frequently Bought Together