Nitrogen receivers (schlenk tubes)
Fournisseur: Lenz Laborglas GmbH & CO.KG
Certificats
À propos de cet article
DURAN®, borosilicate glass, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With 14/23 standard ground sockets/cones and PTFE needle-valve stopcock with tube connector.
Frequently Bought Together