Order Entry
Switzerland
ContactUsLinkComponent
Accessories for safety cutter and knives
Accessories for safety cutter and knives
Catalog # 223-0115
Supplier:  MARTOR ARGENTAX
undefined
Accessories for safety cutter and knives
Catalog # 223-0115
Supplier:  MARTOR ARGENTAX

Specifications

  • Colour:
    Blue
  • Description:
    Small belt holster, plastic, for compact safety knives (SECUNORM MULTISAFE/MULTISET/PROFI/PROFI LIGHT/PROFI25/PROFI40)
  • Seq:
    50
  • Total length:
    104 mm
  • Pk:
    1

Specifications

About this item

This blue plastic holster is ideal for storing and transporting thin cutting tools.

Suitable for: SECUNORM cutters MULTISAFE, MULTISET, PROFI, PROFI LIGHT, PROFI25 and PROFI40.

Weight: 24 g