Order Entry
Switzerland
ContactUsLinkComponent
Acetonitrile : water : trifluoroacetic acid (50 : 47.5 : 2.5) %, LabReady™, Solvent Blend for mass spectrometry, Riedel-de Haën™
Irritant
Health
Flammable
Acetonitrile : water : trifluoroacetic acid (50 : 47.5 : 2.5) %, LabReady™, Solvent Blend for mass spectrometry, Riedel-de Haën™
Catalog # 19182-250ML
CAS Number:  
Irritant
Health
Flammable
Acetonitrile : water : trifluoroacetic acid (50 : 47.5 : 2.5) %, LabReady™, Solvent Blend for mass spectrometry, Riedel-de Haën™
Catalog # 19182-250ML
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

Specification Test Results

  • Assay trifluoroacetic acid:
    2.0 - 3.0 %
  • Water (Karl Fischer):
    47 - 49 %
  • Barium (Ba):
    Max. 5 ppm
  • Calcium (Ca):
    Max. 20 ppm
  • Cadmium (Cd):
    Max. 5 ppm
  • Cobalt (Co):
    Max. 5 ppm
  • Chromium (Cr):
    Max. 5 ppm
  • Copper (Cu):
    Max. 5 ppm
  • Iron (Fe):
    Max. 20 ppm
  • Potassium (K):
    Max. 20 ppm
  • Magnesium (Mg):
    Max. 5 ppm
  • Manganese (Mn):
    Max. 5 ppm
  • Sodium (Na):
    Max. 20 ppm
  • Nickel (Ni):
    Max. 5 ppm
  • Lead (Pb):
    Max. 5 ppm
  • Zinc (Zn):
    Max. 5 ppm

Specification Test Results

About this item

 :
Flash Pt:<10 °C
Storage Temperature:Ambient
UN:1648
ADR:3,II
 :GHS07
 :GHS08
 :GHS02
 :76-05-1