Order Entry
Switzerland
ContactUsLinkComponent
Novec™ 7100 engineered fluid
Novec™ 7100 engineered fluid
Catalog # SHH0002-500ML
CAS Number:  
undefined
Novec™ 7100 engineered fluid
Catalog # SHH0002-500ML
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :250.06 g/mol
CAS Number:163702-08-7;163702-07-6
Formula:C₅H₃OF₉
MDL Number:MFCD28505595