ContactUsLinkComponent
J.T.Baker® BAKERBOND spe™ PCB-N, SPE Columns for Special Applications
J.T.Baker® BAKERBOND spe™ PCB-N, SPE Columns for Special Applications
Catalog # 7524-04
Supplier:  Avantor
J.T.Baker® BAKERBOND spe™ PCB-N, SPE Columns for Special Applications
Catalog # 7524-04
Supplier:  Avantor
Specifications
  • Capacity:
    3 ml
  • Description:
    Column with 500 mg aromatic sulphonic acid (top) and 500 mg silica gel, separated by a PE frit
  • Bed weight:
    1000 mg
  • Phase:
    PCB-N
  • Pk:
    50
Specifications
About this item
This solid phase extraction column is designed for the extraction of PCB’s in oil.