ContactUsLinkComponent
J.T.Baker® BAKERBOND spe™ PAH Aqua, SPE Columns for Special Applications
J.T.Baker® BAKERBOND spe™ PAH Aqua, SPE Columns for Special Applications
Catalog # 7490-08
Supplier:  Avantor
J.T.Baker® BAKERBOND spe™ PAH Aqua, SPE Columns for Special Applications
Catalog # 7490-08
Supplier:  Avantor
Specifications
  • Capacity:
    6 ml
  • Description:
    Column with 500 mg amino (top) and 1000 mg C18, separated by a PE frit
  • Seq:
    02
  • Carbon load:
    1000 mg
  • Bed weight:
    1500 mg
  • Phase:
    PAH Aqua
  • Pk:
    30
Specifications
About this item
These solid phase extraction columns are designed for the extraction and clean-up of Poly Aromatic Hydrocarbons (PAH) from water, including the 16 priority pollutant EPA PAH’s and for several applications using different SPE columns.