Order Entry
Switzerland
ContactUsLinkComponent
J.T.Baker® BAKER INSTRA-ANALYZED®, Zinc Plasma Standard Solution
J.T.Baker® BAKER INSTRA-ANALYZED®, Zinc Plasma Standard Solution
Catalog # 5791.0100
Supplier:  Avantor
CAS Number:  
undefined
J.T.Baker® BAKER INSTRA-ANALYZED®, Zinc Plasma Standard Solution
Catalog # 5791.0100
Supplier:  Avantor
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Bottle
  • Pk:
    100 mL
  • Seq:
    02
  • Element:
    Zinc
  • Symbol:
    Zn

Specifications

About this item

Single element plasma standards are used for accuracy and easy of calibration, along with precise detection of trace impurities.

Certifications: Traceable to NIST.
Caution: 10,000 µg/ml= (1,00% w/v)= 10000 mg/L