Volumetric flasks, class A, glass, amber, DAkkS
Supplier: GLASWARENFABRIK KARL HECHT
Certificates
About this item
Borosilicate glass 3.3, amber, with glass stopper.
- DAkkS calibrated
- Calibrated to contain (TC, In)
- White marks and inscriptions
Certifications: DIN EN ISO 1042, DAkkS and Tolerance certified.
Frequently Bought Together