Visual colour kits, carbon dioxide, MQuant®
Catalog # 1.17179.0001
Anbieter: Merck
CAS-Nummer:
Technische Daten
- Chemical method:Titrimetric with sodium hydroxide
- Pk:0,13 L
- Description:Carbon dioxide
- Seq:01
- Tests per kit:100
- Range:5 - 120 mg/L CO₂
Technische Daten
Über diesen Artikel
Carbon dioxide is an important parameter for the assessment of the surface water quality and of the corrosive behavior of water. The kit is used for water and wastewater treatments following titrimetric method.
The consumption of titration solution is dependent on the concentration of the substance to be determined.
Delivery: Kit includes a bottle of reagent CO2-1 (indicator solution); 2 bottles of reagent CO2-2 (titration solution), a measuring beaker, one graduated test vessel and a card with brief instruction.