ContactUsLinkComponent
VWR®, Measuring Beakers with Handle, PMP TPX
  213-0509
 :
Sustainable Packaging
undefined
VWR®, Measuring Beakers with Handle, PMP TPX
  213-0509
 :
  Sustainable Packaging
Select an Option
5000 ml
Select an Option
06
Select an Option
1
  • Catalog No:
  • 213-0509
  • 213-0508
  • 213-0507
  • 213-0506
  • 213-0505
  • 213-0504
  • Environmentally Preferable:
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
Made of PMP, crystal-clear.
  • With printed graduations
  • Excellent chemical resistance
  • With handle for better usage
  • Withstand temperatures of 0 to 121 °C when used continuously (up to 170 °C for short periods)
  • Resin meets US FDA 21 CFR requirements
Frequently Bought Together