Order Entry
Switzerland
ContactUsLinkComponent
Air compressor, Solaris
Air compressor, Solaris
Catalog # 564-0278
Supplier:  PEAK SCIENTIFIC
undefined
Air compressor, Solaris
Catalog # 564-0278
Supplier:  PEAK SCIENTIFIC

Specifications

  • Description:
    Air compressor, Solaris
  • Seq:
    01
  • Pk:
    1

Specifications

About this item

This air compressor generator has been designed to support the Solaris nitrogen generator for ELSD in laboratories without an external supply of high quality air.

  • Stackable with Solaris nitrogen generator for ELSD and Compact Mass Specs
  • Designed specifically for ELSD and Compact Mass Specs

Solaris has been developed to provide an on-site gas supply of high purity nitrogen for ELSD for laboratories using HPLC or alternatively to supply Compact Mass Spectrometer instruments.

Min./max. operating temperature: 5 to 35 °C

Max. pressure: 8,27 bar

Gas outlet: 1×¼" BSPP

Certifications: Manufactured and tested to highest spec in UK, CE/FCC certified.