Human transferrin (from Orzya sativa)
Catalog # BIRBORB81429-100
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:100 mg
- Source:Orzya sativa
- Species:Human
- Seq:01
- Storage conditions:−20 °C
- Animal-free:Y
- Protein/peptide name:Transferrin
- Purity:95%
- Molecular weight:76 kDa
- Formulation:The protein (1 mg/ml) was lyophilised with no additives
- Tested applications:SDS-PAGE
Specifications
About this item
Recombinant human transferrin produced in plant is non-glycosylated, polypeptide chain containing 679 amino acids.
- Animal-free
- Prevent freeze-thaw cycles
- Well defined and consistent
- Universal iron carrier in cell culture
- Replaces plasma-derived human and bovine transferrin
- Replaces iron chelators
It increases the growth of numerous cell types including hybridoma and primary cell lines. The optimal concentration for cell culture differ with the cell line. The reported effective concentrations for hybridoma and vero cell lines range from 5 to 20 mg/L. Effective concentrations for primary cells range between 5 to 100 mg/L.
One mg of of the protein can bind to approximately micrograms of Fe. Recommended stock concentrations are 5 to 20 mg/ml in PBS, though others can be used as well. Avoid the formation of bubbles when dissolving the protein.