ContactUsLinkComponent
Reference standards for food analysis, FAME mixtures, PUFA, Supelco®
Reference standards for food analysis, FAME mixtures, PUFA, Supelco®
Catalog # 47033.
Supplier:  Merck
CAS Number:  
Reference standards for food analysis, FAME mixtures, PUFA, Supelco®
Catalog # 47033.
Supplier:  Merck
CAS Number:  
Specifications
  • Pk:
    100 mg
  • Description:
    PUFA No.1 (marine source)
  • Seq:
    01
Specifications
About this item
Complex qualitative polyunsaturated fatty acid (PUFA) methyl esters standard mixtures, extracted from natural materials.