ContactUsLinkComponent
Alkaline detergents, powder, LaboClean F, neodisher®
Alkaline detergents, powder, LaboClean F, neodisher®
  148-3139
undefined
Alkaline detergents, powder, LaboClean F, neodisher®
  148-3139
  • Description:
    LaboClean F, Alkaline detergent
  • Pk:
    4
LaboClean F detergent is composed of alkaline components and dispersants. Ideal for the removal of grease, oil, paraffin, resin, organic dyes and residues of indelible pen.
  • High complexing and dispersive action
  • Used for intensive cleaning
  • Free of phosphates and surfactants
It is not suitable for non ferrous and light metals.