ContactUsLinkComponent
Disintegration testing accessories, QLA
Disintegration testing accessories, QLA
Catalog # QLAAGLA905-DB
undefined
Disintegration testing accessories, QLA
Catalog # QLAAGLA905-DB
Specifications
  • Description:
    Disintegration beaker with flat top, clear glass, 900 ml
  • Seq:
    08
  • Pk:
    1
Specifications
About this item
Disintegration testing accessories and glassware for Agilent/VanKel disintegration testers.
  • Fluted disks can be used with any brand of disintegration tester
Certifications: Complies to USP.