Anti-Dio-1 Rabbit Polyclonal Antibody [clone: S186]
Catalog # BWRLBS2533
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Dio-1
- Clonality:Polyclonal
- Clone:S186
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 154-198 of Human Dio-1.
Dio-1 (S186) pAb detects endogenous levels of Dio-1 protein.
Type: Primary
Antigen: Dio-1
Clonality: Polyclonal
Clone: S186
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat