Face light M8 with light guide
Catalog # ZEIS4355080000000
Supplier: ZEISS
Specifications
- Description:Diffuse face light M8 (D) radiating surface 78×30 mm with light guide 8/1000 mm and M6/M8 screw adapter
- Environmentally Preferable:
- Pk:1
Specifications
About this item
Diffuse face light M8 with light guide (D) radiating surface 78×30 mm, outer dimensions 140×58×22 mm, incl. light guide 8/1000 mm and M6/M8 screw adapter.