Order Entry
Switzerland
ContactUsLinkComponent
 
Electrode extension cables, Orion™
  662-0769
Electrode extension cables, Orion™
  662-0769

 

  • Description:
    Electrode extension cable, 15 ft. (4,57 m), BNC connector
  • Seq:
    02
  • Pk:
    1

 

 

These cables can be used with a variety of electrodes.

  • Extend the length of an electrode cable by 15 ft. (4,57 m)
  • Different connectors available