Orders LinkContactUsLinkComponent
Electrode Adapter Cables
Electrode Adapter Cables
Supplier:   Thermo Orion
Electrode Adapter Cables
Supplier:  Thermo Orion
Certificates
Electrode Adapter Cables
2 Options Available Below
Product Details & Documents
These cables can be used to convert an electrode with one connector to the required connector of another meter.
For use with meters that have BNC, US standard, certain LEMO, British, E DIN, Radiometer, 2 mm reference pins or just a stripped lead inputs.
Electrodes must have a US standard, screw cap, Karl Fisher, or BNC connectors.
Product Family Options
Product Information
  • Availability
    Price
  • 662-1535
    Adapter, BNC meter input + screw cap electrode connector; detachable cable to BNC connector
    Adapter, BNC meter input + screw cap electrode connector; detachable cable to BNC connector
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 112.00
    SFr. 112.00
    1 * 1 ST
       SFr. 112.00
  • ORIO91USRF
    Adapter, 2 mm pin tip meter input + screw cap electrode connector; detachable cable to 2,5 mm pin tip connector
    Adapter, 2 mm pin tip meter input + screw cap electrode connector; detachable cable to 2,5 mm pin tip connector
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 288.00
    SFr. 288.00
    1 * 1 ST
       SFr. 288.00