ContactUsLinkComponent
Hard-sided field case for portable meters, Orion™ Star A series
Hard-sided field case for portable meters, Orion™ Star A series
  ORIOSTARA-CS
undefined
Hard-sided field case for portable meters, Orion™ Star A series
  ORIOSTARA-CS
  • For:
    Orion Star A120, A220 + A320 series
  • Description:
    Hard-sided field case for portable meters, holds meter (with or without armor), portable electrode holders, extra batteries, solutions, pH electrode in storage sleeve, conductivity probe and dissolved oxygen probe in storage sleeve
  • Seq:
    105
  • Pk:
    1
This hard-sided field case can be used to protect, transport and store measuring equipment in the field.
The field case holds Star A series portable meter (with or without armor), portable electrode holders, extra batteries, solutions, pH electrode in storage sleeve, conductivity probe and dissolved oxygen probe in storage sleeve. The electrodes and probes can remain connected to meter while in the case plus the internal separating door can be removed for cable management.
For use with: Thermo Scientific™ Orion™ Star A series portable meters (Star A320, Star A220 and Star A120 series)