ContactUsLinkComponent
Protective armor for portable meters, Orion™ Star A series
Catalog # ORIOSTARA-AR
Supplier:  Thermo Orion
undefined
Protective armor for portable meters, Orion™ Star A series
Catalog # ORIOSTARA-AR
Supplier:  Thermo Orion
Specifications
  • Description:
    Protective armor for portable meters, incl. hand strap and built-in stand, pH electrode storage sleeve, pH electrode holder, conductivity and dissolved oxygen sensor holder
  • Seq:
    100
  • Pk:
    1
Specifications
About this item
This portable meter armor adds protection around the meter screen and includes a hand or belt strap that can be converted to built-in stand, pH electrode holder, conductivity and DO probe holder and pH electrode storage sleeve.
For use with Thermo Scientific™ Orion™ Star A series portable meters (Star A320, Star A220 and Star A120 series).
Delivery: Supplied with hand strap and built-in stand, pH electrode storage sleeve, pH electrode holder, and conductivity and dissolved oxygen sensor holder.
Replacement electrode holders
Product Information