Capillary columns, HT5
Supplier: Trajan Scientific and Medical
Certificates
About this item
Unique high temperature phase suited for simulated distillation and other petroleum applications.
5% phenyl polycarborane-siloxane.
Maximum temperature: polyimide 380/400 °C; aluminium 460/480 °C
Suitable replacement for: MXT-1 Sim Dist, HT-Sim, DistCB, MXT-500.
Specifications
- Catalog No:
- 568-0188
- 568-0199
- 568-0193
- 568-0185
- 568-0189
- 568-0184
- 568-0194
- 568-0183
- 568-0195
- 568-0182
- Temperature limit:
- 10 to 380/400 °C
- 10 to 380/400 °C
- 10 to 380/400 °C
- 10 to 380/400 °C
- 10 to 380/400 °C
- 10 to 380/400 °C
- 10 to 380/400 °C
- 10 to 380/400 °C
- 10 to 380/400 °C
- 10 to 380/400 °C