Bag openers for Stomacher® bags
Nome del fornitore: Seward (FermionX)
Certificati
Informazioni su questo articolo
Made from high grade plastic polymer.
- Compatible with Stomacher® 80 bags
- Stable on bench-top or scales
- Easily cleaned with most lab disinfectants
These bag openers hold open bags for easy addition of sample without risk of instrument contact on the bag.
Frequently Bought Together