Racks for Stomacher® bags
Supplier: Seward (FermionX)
Certificates
About this item
Made from high grade plastic polymer.
- Perfect for loading closure bags preenrichments into the incubator
- Variety of capacities for each Stomacher® machine
- Easily dismantled and cleaned with most lab disinfectants (not autoclavable)
Bag racks provide the perfect storage for Stomacher® bag sample handling, helping to provide a clean and productive working environment.
Frequently Bought Together