VWR®, Reagent Bottles, Narrow Neck, with Standard Ground Stoppers
Nome del fornitore: VWR Collection
Certificati
Informazioni su questo articolo
Borosilicate 3.3 glass according to ISO 4796.
- High resistance to chemicals and thermal shock
- Available in capacities from 10 to 2000 ml
- Hollow glass stopper and ground joint
These reagent bottles have a precise ground neck to ensure a water- and air-tight connection between the bottle and the glass stopper. All components are made from borosilicate 3.3 glass and are, therefore, ideal for the storage of aggressive and hot media.
Informazioni correlate
Frequently Bought Together