Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Agarose
Agarose, QA-Agarose™, low melting
Agarose, QA-Agarose™, low melting
Agarose, QA-Agarose™, low melting
Catalog # ICNA11AGAL0050
Supplier:
MP Biomedicals
CAS Number:
Expand image
Agarose, QA-Agarose™, low melting
Catalog # ICNA11AGAL0050
Supplier:
MP Biomedicals
CAS Number:
Specifications
Pk:
50 g
Seq:
3368
Specifications
About this item
Product Details
:
Storage Temperature:
Ambient
CAS Number:
9012-36-6
MDL Number:
MFCD00081294
Product Details