Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Agarose, basic first | Avantor

Agarose, basic first

Supplier:   MP Biomedicals
Agarose, basic first
Supplier:  MP Biomedicals
Certificates
Agarose, basic first
Agarose, basic first

2 Options Available Below

Product Details & Documents

Basic agarose premier is ideal for routine rapid separation of DNA and RNA fragments as well as PCR products, the preparation of plasmids, and for screening, cloning and blotting techniques.

Agarose is a purified linear galactan hydrocolloid isolated from agar or agar-bearing marine algae.

  • CAS Number:9012-36-6
  • MDL Number:MFCD00081294
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 96.90 SFr.
    96.90 SFr.
    1 * 100 g
       96.90 SFr.

  • 500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 459.00 SFr.
    459.00 SFr.
    1 * 500 g
       459.00 SFr.