Order Entry
Switzerland
ContactUsLinkComponent
Tritirachium album proteinase K, MP Biomedicals
Tritirachium album proteinase K, MP Biomedicals
Supplier:  MP Biomedicals
Certificates
Tritirachium album proteinase K, MP Biomedicals
Supplier:  MP Biomedicals
Certificates

Specifications

  • CAS No.:
    39450-01-6
  • Species:
    Tritirachium album
  • Storage conditions:
    +4 °C, dessicate
  • MDL No.:
    MFCD00132129
  • Enzyme name:
    Proteinase K
  • Enzyme activity:
    >3 Manson U/Mg
  • Grade:
    Molecular biology reagent

Specifications

Related Information

Frequently Bought Together