Bacillus cereus beta-Lactamase, MP Biomedicals
ICNA0219003401
:
- Pk:1 Vial
- CAS No.:9073-60-3
- Species:Bacillus cereus
- Storage conditions:+4 °C
- MDL No.:MFCD00131819
- Enzyme name:Β-Lactamase
- Molecular weight:32000
Activity: Each vial contains 500 units β-Lactamase I and 50 units β-Lactamase II Unit Definition: One unit is defined as the amount of enzyme which will hydrolyse 1,0 micromole of benzyl penicillin and 1,0 micromole of cephalosporin C per minute at 25 °C in the presence of EDTA pH 7,0.
E.C. 3.5.2.6
Lyophilised solid