Myrothecium sp. Bilirubin oxidase, MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
- Lyophilised powder
- Activity: ≥0,8 u/mg solid
- Unit definition: one unit is the amount of enzyme that oxidises one micromole of bilirubin per minute at 37 °C, pH 8,4.
Specifications
- CAS No.:80619-01-8
- Species:Myrothecium sp.
- Storage conditions:+4 °C
- Enzyme name:Bilirubin Oxidase
Specifications
Frequently Bought Together