Sorbitol dehydrogenase, MP Biomedicals
Catalog # ICNA0215664205
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:5 mg
- CAS No.:9028-21-1
- Storage conditions:−20 °C
- Enzyme name:Sorbitol Dehydrogenase
Specifications
About this item
Sorbitol dehydrogenase has been used in a study to investigate osmotic stress induced oxidative damage as a possible mechanism of cataract formation in diabetes.
Sorbitol dehydrogenase is an oxidoreductase. Sorbitol dehydrogenase is an enzyme in carbohydrate metabolism converting fructose into Sorbitol.
Biochem/physiol actions: activity of sorbitol dehydrogenase increases in diabetic animals.
Unit definition: one unit will convert 1,0 µmole of D-fructose to D-sorbitol per minute at pH 7,6 at 25 °C.