Pig Pepsin (from Gastric mucosa), MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
Stable powder 3X
Specifications
- CAS No.:9001-75-6
- Source:Gastric mucosa
- Species:Pig
- Storage conditions:Ambient
- Form:Off-white lyophilized powder
- Enzyme name:Pepsin
- Enzyme activity:1:10 000
- Molecular weight:~35000
- Formulation:Lyophilised powder
- pH:2.5 - 4.5
Specifications
Frequently Bought Together