Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Aluminium oxide basic, MP Ecochrom™ Super I

Aluminium oxide basic, MP Ecochrom™ Super I

500 g
Catalog # ICNA0204568
Supplier:   MP Biomedicals
Aluminium oxide basic, MP Ecochrom™ Super I
Catalog # ICNA0204568
Supplier:  MP Biomedicals
CAS Number:  1344-28-1
Aluminium oxide basic, MP Ecochrom™ Super I
Aluminium oxide basic, MP Ecochrom™ Super I

500 g

CAS Number:  1344-28-1
Price
SFr. 58.20
Log in for availability
1 * 500 g
Product Details & Documents

MP EcoChrom™ Alumina B, Activity Super I available as basic alumina is commonly used for purification processes of all kinds, including sample preparation for pesticides, and environmental analysis, solvent purification and all kinds of chromatography, requiring the highest precision and reproducibility.

  • Specific Surface Area: 200 m2/g
  • Particle Size: 63-200 µm
  • Weight: 0.8 g/ml
  • pH: 10

pH: 10

  • CAS Number:1344-28-1
  • Formula:Al₂O₃
  • MDL Number:MFCD00003424
  • Molecular Weight:101.96 g/mol
  • Boiling Pt:∼2980 °C
  • Melting Pt:∼. 2072 °C
  • Density:3.94 g/cm³ (20 °C)
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.