ContactUsLinkComponent
Cutlery, multi-purpose, Corning® Gosselin™
Cutlery, multi-purpose, Corning® Gosselin™
Catalog # GOSSCFM-01
Supplier:  Corning
   Low Manufacturing Impact
undefined
Cutlery, multi-purpose, Corning® Gosselin™
Catalog # GOSSCFM-01
Supplier:  Corning
   Low Manufacturing Impact
Specifications
  • Length:
    180 mm
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    RECs, ISO 14001 - Borre
  • Pk:
    500
Specifications
About this item
Multi-purpose cutlery made from white PS. For sample collection and processing in food, pharmaceutical or cosmetic industries.
  • Food contact approved: Raw material is compliant with regulation EC N°1935/2004
  • Sterility assurance level based on ISO 11137: STERILE SAL 10⁻³
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904