Orders LinkContactUsLinkComponent
Tray, Rotilabo®
Tray, Rotilabo®
Marketsource
Supplier:   Roth Carl
Tray, Rotilabo®
Supplier:  Roth Carl
Marketsource
Certificates
Tray, Rotilabo®
4 Options Available Below
Product Details & Documents
Dishes, made of stainless steel, chrome-nickel plated.
  • With folding handles on two sides
  • Very good temperature resistance
  • Easy to clean
  • Autoclavable
Suitable for food applications.
Product Family Options
Product Information
  • W×D×H
    Availability
    Price
  • 365×270×75 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 122.00
    SFr. 122.00
    1 * 1 ST
       SFr. 122.00
    Specifications:
    Marketsource
    Marketsource
  • 470×325×90 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 198.00
    SFr. 198.00
    1 * 1 ST
       SFr. 198.00
    Specifications:
    Marketsource
    Marketsource
  • 520×325×90 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 244.80
    SFr. 244.80
    1 * 1 ST
       SFr. 244.80
    Specifications:
    Marketsource
    Marketsource
  • 630×450×90 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 381.60
    SFr. 381.60
    1 * 1 ST
       SFr. 381.60
    Specifications:
    Marketsource
    Marketsource