Industrial overalls, AlphaTec® 1800 Standard, model 111
Supplier: Ansell
Certificates
About this item
These overalls are made from a lightweight, breathable microporous PE laminate with a non woven inner layer. This fabric is particularly suited to warmer working environments and provides a good barrier to low hazard liquid spray and fine particulates.
- Three-piece hood, elasticated wrists, waist and ankles
- Two-way front zipper with re-sealable storm flap
- Bound seams enhance overall robustness and barrier protection
- Moisture vapour permeable (‘breathable’) to help reduce the risk of heat stress
- Silicone-free: Critical in spray painting applications
- Low linting: Reduces the risk of fibre contamination in some critical areas
- Optimised body fit improves wearer comfort and safety
Typical applications include: General maintenance, paint spraying, powder handling.
Weight: 53 g/m²
Certifications: Type 5-B/6-B, EN 1149-5; Cat. III
Specifications
- Catalog No:
- ANSEWH18B-00111-07
- ANSEWH18B-00111-06
- ANSEWH18B-00111-05
- ANSEWH18B-00111-04
- ANSEWH18B-00111-03
- ANSEWH18B-00111-02
- Model:
- 111
- 111
- 111
- 111
- 111
- 111
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Anti-static:
- +
- +
- +
- +
- +
- +
- Seam type:
- Bound
- Bound
- Bound
- Bound
- Bound
- Bound
- Garment application:
- Chemical protection, General Purpose, Industrial
- Chemical protection, General Purpose, Industrial
- Chemical protection, General Purpose, Industrial
- Chemical protection, General Purpose, Industrial
- Chemical protection, General Purpose, Industrial
- Chemical protection, General Purpose, Industrial
- Weight:
- 53 g/m²
- 53 g/m²
- 53 g/m²
- 53 g/m²
- 53 g/m²
- 53 g/m²
Specifications
Frequently Bought Together