Industrial overalls, AlphaTec® 2300 COMFORT, model 129
Supplier: Ansell
Certificates
About this item
These overalls have been engineered for workers involved in composites manufacturing and related industries.
- Three-piece hood - ensures a good fit with half and full face respirators
- Elasticated hood, wrists, waist and ankles
- Finger loops, prevent sleeve movement whilst working above your head
- Two-way front zipper with resealable storm flap
- Breathable SMS back panel
- Silicone-free: Critical in spray painting applications
- Fabric construction ensures that there is no risk of delamination should a wearer make contact with a tacky surface
The critical areas to the front of the garment (including the hood, arms and legs) are made of a PE barrier coating with a bicomponent non woven inner layer. The fabric provides an excellent barrier to resins, fibres and many other hazards associated with composites manufacturing.
The back panel is made from a fabric which is air and water vapour permeable whilst providing a good barrier to fibres and particulates. This panel allows airflow around the suit, increasing wearer comfort.
Typical applications include: Composites manufacturing, pharmaceutical industries, general maintenance.
Weight: 62 g/m²
Certifications: Type 5/6, EN 1073-2; EN 1149-5; Cat. III.
Specifications
- Catalog No:
- ANSEWY23B-00129-06
- ANSEWY23B-00129-05
- ANSEWY23B-00129-04
- ANSEWY23B-00129-03
- ANSEWY23B-00129-07
- Model:
- 129
- 129
- 129
- 129
- 129
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Anti-static:
- +
- +
- +
- +
- +
- Seam type:
- Bound
- Bound
- Bound
- Bound
- Bound
- Garment application:
- Chemical protection, General Purpose, Industrial
- Chemical protection, General Purpose, Industrial
- Chemical protection, General Purpose, Industrial
- Chemical protection, General Purpose, Industrial
- Chemical protection, General Purpose, Industrial
- Weight:
- 62 g/m²
- 62 g/m²
- 62 g/m²
- 62 g/m²
- 62 g/m²
Specifications
Frequently Bought Together