ContactUsLinkComponent
Industrial overalls, AlphaTec® 1500, model 138
Industrial overalls, AlphaTec® 1500, model 138
Supplier:  Ansell
Certificates
undefined
Industrial overalls, AlphaTec® 1500, model 138
Supplier:  Ansell
Specifications
  • Catalog No:
  • ANSEWH15S-00138-06
  • ANSEWH15S-00138-05
  • ANSEWH15S-00138-04
  • ANSEWH15S-00138-03
  • ANSEWH15S-00138-02
  • ANSENV15S-00138-07
  • ANSENV15S-00138-06
  • ANSENV15S-00138-05
  • ANSENV15S-00138-04
  • ANSENV15S-00138-03
  • ANSEWH15S-00138-07
  • Model:
  • 138
  • 138
  • 138
  • 138
  • 138
  • 138
  • 138
  • 138
  • 138
  • 138
  • 138
  • Closure type:
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Seam type:
  • Taped
  • Taped
  • Taped
  • Taped
  • Taped
  • Taped
  • Taped
  • Taped
  • Taped
  • Taped
  • Taped
  • Garment application:
  • General Purpose, Industrial
  • General Purpose, Industrial
  • General Purpose, Industrial
  • General Purpose, Industrial
  • General Purpose, Industrial
  • General Purpose, Industrial
  • General Purpose, Industrial
  • General Purpose, Industrial
  • General Purpose, Industrial
  • General Purpose, Industrial
  • General Purpose, Industrial
  • Weight:
  • 48 / 50 g/m²
  • 48 / 50 g/m²
  • 48 / 50 g/m²
  • 48 / 50 g/m²
  • 48 / 50 g/m²
  • 48 / 50 g/m²
  • 48 / 50 g/m²
  • 48 / 50 g/m²
  • 48 / 50 g/m²
  • 48 / 50 g/m²
  • 48 / 50 g/m²