Chemical resistant overalls, with integrated socks, Microchem® 4000, model 122
Supplier: Ansell
Certificates
About this item
These overalls are made of multi-layer barrier fabric (external film barrier, non woven inner layer, core high barrier film). They are designed to provide an exceptional barrier against organic and inorganic chemicals and biological agents.
- Double zip system helps ensure a liquid tight seal without the need for additional taping
- Double cuffs with knitted inner cuff
- Elasticated 2-piece hood, outer cuffs, waist and boot overflaps
- Integrated socks with boot overflap
- Ultrasonically welded and taped seams
- Textile-like inner improves wearer acceptance
- Permeation tested against over 180 chemicals, including chemical warfare agents
Typical applications include: Chemical handling, transportation, oil-based mud protection, HAZMAT emergency response, pharmaceutical, mining and agriculture.
Certifications: Type 3/4/5, EN 14126, EN 1073-2, EN 1149-5; Cat. III
Suitable for Ex-Zones.
Suitable for Ex-Zones.
Specifications
- Catalog No:
- ANSEGR40T-00122-07
- ANSEGR40T-00122-06
- ANSEGR40T-00122-05
- ANSEGR40T-00122-04
- ANSEGR40T-00122-03
- ANSEGR40T-00122-02
- Model:
- 122
- 122
- 122
- 122
- 122
- 122
- Anti-static:
- +
- +
- +
- +
- +
- +
- Garment application:
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial