Order Entry
Switzerland
ContactUsLinkComponent
 
Adenosine-5ʹ-diphosphate, potassium salt dihydrate ≥98% (by enzymatic purity), Millipore®
Catalog # 117105-1
CAS Number:  
Adenosine-5ʹ-diphosphate, potassium salt dihydrate ≥98% (by enzymatic purity), Millipore®
Catalog # 117105-1
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Form:
    White solid
  • Purity:
    ≥98% by enzymatic assay (dry basis)
  • Contaminants:
    AMP: ≤1% and ATP: ≤0,2% by enzymatic assay
  • Solubility:
    H2O (50 mg/ml)
  • Storage:
    2 to 8 °C; Hygroscopic
  • Stability:
    Following reconstitution, aliquot and freeze (−20 °C). Stock solutions are stable for up to 6 months at −20 °C.

Specification Test Results

About this item

 :
 :501.3 g/mol
Storage Temperature:Refrigerator
CAS Number:72698-48-1
Formula:C₁₀H₁₄N₅O₁₀P₂K·2H₂O