ContactUsLinkComponent
Filling solution for ISE electrodes, sodium
Filling solution for ISE electrodes, sodium
Katalognr. HACH2965126
Supplier:  Hach
CAS Number:  
undefined
Filling solution for ISE electrodes, sodium
Katalognr. HACH2965126
Supplier:  Hach
CAS Number:  
Specifications
  • Pk:
    59 mL
  • Description:
    Electrode filling solution, 0,02 M NH4Cl, for use with ISENA381 IntelliCAL™ electrode, 59 ml bottle, with dispenser cap
Specifications
About this item
Internal filling solution for use with ISENA381 IntelliCAL™ Ultra sodium ion selective electrodes.